siRNA Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290699.1 | NP_001277628.1 | 10 | 2773 | ||
| NM_133817.3 | NP_598578.1 | 12 | 3011 | |||
| GenBank mRNA | AK030088.1 | - | 12 | 2844 | ||
| BC024435.1 | - | 3 | 686 | |||
| BC026587.1 | - | 3 | 1189 | |||
| BC043685.1 | - | 3 | 1261 | |||
| BC062154.1 | - | 10 | 2563 | 6331597 | ||

Zfp451
Genomic Map