siRNA Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178405.3 | NP_848492.1 | 21 | 3057 | ||
| GenBank mRNA | AK129212.1 | - | 21 | 2970 | ||
| AK137602.1 | - | 21 | 2993 | |||
| AK147331.1 | - | 21 | 2989 | |||
| AK148775.1 | - | 20 | 3179 | |||
| AK161840.1 | - | 21 | 2989 | |||
| AK164509.1 | - | 21 | 2993 | |||
| AK168703.1 | - | 21 | 2989 | |||
| BC025807.1 | - | 10 | 1285 | |||
| BC036127.1 | - | 21 | 2981 | 4955003 | ||
| BC041774.1 | - | 18 | 2707 | |||

Atp1a2
Genomic Map