siRNA Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178405.3 | NP_848492.1 | 18, 19 | 2725 | ||
| GenBank mRNA | AK129212.1 | - | 18, 19 | 2638 | ||
| AK137602.1 | - | 18, 19 | 2661 | |||
| AK147331.1 | - | 18, 19 | 2657 | |||
| AK161840.1 | - | 18, 19 | 2657 | |||
| AK164509.1 | - | 18, 19 | 2661 | |||
| AK168703.1 | - | 18, 19 | 2657 | |||
| BC025807.1 | - | 7, 8 | 953 | |||
| BC036127.1 | - | 18, 19 | 2649 | 4955003 | ||
| BC041774.1 | - | 15, 16 | 2375 | |||

Atp1a2
Genomic Map