siRNA Details
| Gene Symbol: Mfsd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133829.2 | NP_598590.2 | 5, 6 | 2242 | ||
| NM_178081.4 | NP_835182.1 | 5, 6 | 2242 | |||
| GenBank mRNA | AB247936.1 | - | 4, 5 | 1857 | ||
| AK035995.1 | - | 5, 6 | 2243 | |||
| AK046866.1 | - | 5, 6 | 2386 | |||
| BC017534.1 | - | 4, 5 | 1351 | 4488390 | ||
| BC029324.1 | - | 4, 5 | 1599 | |||

Mfsd6
Genomic Map