siRNA Details
| Gene Symbol: Obsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178884.5 | NP_849215.3 | 12 | 4167 | ||
| GenBank mRNA | AK029863.1 | - | 12 | 3956 | ||
| AK048144.1 | - | 12 | 3359 | |||
| AK122340.1 | - | 6 | 2286 | |||
| BC025848.1 | - | 1 | 106 | |||
| BC056205.1 | - | 10 | 2227 | |||

Obsl1
Genomic Map