siRNA Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133835.2 | NP_598596.2 | 9 | 1264 | ||
| GenBank mRNA | AK157857.1 | - | 9 | 1098 | ||
| AK200854.1 | - | 1 | 42 | |||
| AK205743.1 | - | 1 | 42 | |||
| BC021811.1 | - | 9 | 1215 | 3662474 | ||

Ubac1
Genomic Map