siRNA Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133837.4 | NP_598598.1 | 6 | 438 | ||
| GenBank mRNA | AK090122.1 | - | 6 | 439 | ||
| AK141463.1 | - | 6 | 426 | |||
| AK159208.1 | - | 6 | 459 | |||
| AK161246.1 | - | 6 | 425 | |||
| BC019753.1 | - | 6 | 413 | 5008314 | ||
| BC022972.1 | - | 6 | 427 | 5368775 | ||
| BC023821.1 | - | 6 | 430 | 5354711 | ||
| BC024787.1 | - | 6 | 425 | 5359215 | ||

Cdc123
Genomic Map