siRNA Details
| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080548.1 | NP_001074017.1 | 15 | 2241 | ||
| NM_181399.3 | NP_852064.2 | 14 | 2080 | |||
| GenBank mRNA | AK084045.1 | - | 15 | 2066 | ||
| AK084524.1 | - | 1 | 382 | |||
| AK129037.1 | - | 15 | 2093 | |||
| AK133119.1 | - | 6 | 1320 | |||
| AK155073.1 | - | 15 | 2242 | |||
| BC051184.1 | - | 14 | 2083 | |||

Usp6nl
Genomic Map