siRNA Details
| Gene Symbol: Lpcat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207206.2 | NP_997089.1 | 3 | 496 | ||
| GenBank mRNA | BC046276.1 | - | 2 | 246 | ||
| BC068131.1 | - | 3 | 496 | 6414152 | ||
| BC080829.1 | - | 3 | 463 | 30435722 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpcat4| Gene Symbol: Lpcat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207206.2 | NP_997089.1 | 3 | 496 | ||
| GenBank mRNA | BC046276.1 | - | 2 | 246 | ||
| BC068131.1 | - | 3 | 496 | 6414152 | ||
| BC080829.1 | - | 3 | 463 | 30435722 | ||