siRNA Details
| Gene Symbol: Pomt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145145.1 | NP_660127.1 | 3, 4 | 312 | ||
| GenBank mRNA | AK053326.1 | - | 3, 4 | 324 | ||
| AK053889.1 | - | 3, 4 | 311 | |||
| AK134770.1 | - | 3, 4 | 356 | |||
| AK153984.1 | - | 3, 4 | 411 | |||
| AK200298.1 | - | 3, 4 | 293 | |||
| AY007238.1 | - | 3, 4 | 352 | |||
| AY494857.1 | - | 3, 4 | 372 | |||
| BC027325.1 | - | 3, 4 | 312 | 3981366 | ||

Pomt1
Genomic Map