siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290733.1 | NP_001277662.1 | 25 | 3296 | ||
| NM_001290734.1 | NP_001277663.1 | 23 | 2963 | |||
| NM_145525.3 | NP_663500.2 | 25 | 3080 | |||
| GenBank mRNA | AK037703.1 | - | 23 | 2914 | ||
| AK044411.1 | - | 25 | 3267 | |||
| AK220498.1 | - | 25 | 3027 | |||
| BC022908.1 | - | 18 | 2140 | 5043261 | ||

Osbpl6
Genomic Map