siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290733.1 | NP_001277662.1 | 19 | 2524 | ||
| NM_001290734.1 | NP_001277663.1 | 17 | 2191 | |||
| NM_145525.3 | NP_663500.2 | 19 | 2308 | |||
| GenBank mRNA | AK037703.1 | - | 17 | 2142 | ||
| AK044411.1 | - | 19 | 2495 | |||
| AK220498.1 | - | 19 | 2255 | |||
| BC022908.1 | - | 12 | 1368 | 5043261 | ||

Osbpl6
Genomic Map