siRNA Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207298.2 | NP_997181.1 | 7, 8 | 997 | ||
| GenBank mRNA | AK155384.1 | - | 7, 8 | 993 | ||
| BC052176.1 | - | 7, 8 | 958 | |||
| BC138847.1 | - | 7, 8 | 976 | 8861869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cercam| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207298.2 | NP_997181.1 | 7, 8 | 997 | ||
| GenBank mRNA | AK155384.1 | - | 7, 8 | 993 | ||
| BC052176.1 | - | 7, 8 | 958 | |||
| BC138847.1 | - | 7, 8 | 976 | 8861869 | ||