siRNA Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175300.4 | NP_780509.2 | 13 | 2399 | ||
| GenBank mRNA | AK033784.1 | - | 13 | 2367 | ||
| AK129351.1 | - | 13 | 2352 | |||
| AK154465.1 | - | 13 | 2353 | |||
| AK170676.1 | - | 13 | 2365 | |||
| AK218217.1 | - | 2 | 143 | |||
| BC027351.1 | - | 13 | 2347 | |||
| BC038267.1 | - | 12 | 1710 | |||
| BC065421.1 | - | 4 | 512 | |||
| BC096381.1 | - | 12 | 2592 | |||
| BC137583.1 | - | 13 | 2367 | 8860602 | ||

Anapc2
Genomic Map