siRNA Details
| Gene Symbol: Wdr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181400.3 | NP_852065.2 | 11, 12 | 2431 | ||
| GenBank mRNA | AK028113.1 | - | 5, 6 | 822 | ||
| BC040337.1 | - | 11, 12 | 2292 | 3586908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr47| Gene Symbol: Wdr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181400.3 | NP_852065.2 | 11, 12 | 2431 | ||
| GenBank mRNA | AK028113.1 | - | 5, 6 | 822 | ||
| BC040337.1 | - | 11, 12 | 2292 | 3586908 | ||