siRNA Details
| Gene Symbol: Usp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133857.3 | NP_598618.1 | 10, 11 | 1827 | ||
| GenBank mRNA | AK036876.1 | - | 10, 11 | 1828 | ||
| AK040332.1 | - | 11, 12 | 1723 | |||
| AK140453.1 | - | 10, 11 | 1871 | |||
| BC096383.1 | - | 10, 11 | 1828 | |||
| BC132339.1 | - | 9, 10 | 1560 | 40130616 | ||
| BC145707.1 | - | 9, 10 | 1560 | 40131010 | ||

Usp53
Genomic Map