siRNA Details
| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170778.2 | NP_740748.1 | 20 | 2654 | ||
| GenBank mRNA | AK054474.1 | - | 1 | 80 | ||
| BC028831.1 | - | 2 | 198 | |||
| BC039699.1 | - | 20 | 2655 | 4235971 | ||
| BC042543.1 | - | 16 | 2119 | |||
| BC044730.2 | - | 8 | 1017 | |||

Dpyd
Genomic Map