siRNA Details
| Gene Symbol: Kcnc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145922.2 | NP_666034.1 | 2 | 1160 | ||
| GenBank mRNA | AK158073.1 | - | 2 | 560 | ||
| BC024837.1 | - | 2 | 1160 | 5358950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnc4| Gene Symbol: Kcnc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145922.2 | NP_666034.1 | 2 | 1160 | ||
| GenBank mRNA | AK158073.1 | - | 2 | 560 | ||
| BC024837.1 | - | 2 | 1160 | 5358950 | ||