siRNA Details
| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153570.2 | NP_705798.2 | 15 | 1569 | ||
| GenBank mRNA | AK052601.1 | - | 15 | 1569 | ||
| AK088355.1 | - | 15 | 1567 | |||
| AK090024.1 | - | 15 | 1581 | |||
| AK146006.1 | - | 15 | 1565 | |||
| AK172310.1 | - | 15 | 1548 | |||
| BC024616.1 | - | 15 | 1559 | 4218310 | ||

Noc4l
Genomic Map