siRNA Details
| Gene Symbol: Prrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101443.1 | NP_001094913.1 | 2 | 626 | ||
| GenBank mRNA | AK052319.1 | - | 2 | 628 | ||
| BC140954.1 | - | 2 | 622 | 9055566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrt4| Gene Symbol: Prrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101443.1 | NP_001094913.1 | 2 | 626 | ||
| GenBank mRNA | AK052319.1 | - | 2 | 628 | ||
| BC140954.1 | - | 2 | 622 | 9055566 | ||