siRNA Details
| Gene Symbol: C230052I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178643.5 | NP_848758.1 | 2 | 648 | ||
| GenBank mRNA | AK031566.1 | - | 2 | 689 | ||
| AK040248.1 | - | 2 | 674 | |||
| AK082457.1 | - | 2 | 649 | |||
| AK153918.1 | - | 2 | 595 | |||
| AK166481.1 | - | 2 | 695 | |||
| BC096687.1 | - | 2 | 500 | 1433264 | ||

C230052I12Rik
Genomic Map