siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159591.1 | NP_001153063.1 | 6 | 790 | ||
| NM_134002.2 | NP_598763.1 | 5 | 967 | |||
| GenBank mRNA | AK077076.1 | - | 6 | 770 | ||
| AK132871.1 | - | 6 | 791 | |||
| AK152413.1 | - | 5 | 966 | |||
| BC004839.1 | - | 5 | 909 | 2648652 | ||
| BC063083.1 | - | 3 | 2091 | |||

Csnk1g2
Genomic Map