siRNA Details
| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134021.2 | NP_598782.1 | 5 | 592 | ||
| GenBank mRNA | AK154004.1 | - | 5 | 532 | ||
| AK154443.1 | - | 5 | 353 | |||
| AK164667.1 | - | 5 | 593 | |||
| BC010785.1 | - | 5 | 531 | 4210325 | ||
| BC026564.1 | - | 5 | 559 | 4971745 | ||
| CK625276.1 | - | 5 | 401 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK029743.1 | - | 954 | Chr. - - [-] on Build | ||

Pnpo
Genomic Map