siRNA Details
| Gene Symbol: Tbc1d10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134023.1 | NP_598784.1 | 1 | 226 | ||
| GenBank mRNA | AK140872.1 | - | 1 | 238 | ||
| AK153677.1 | - | 208 | ||||
| AK220243.1 | - | 237 | ||||
| BC018300.1 | - | 1 | 226 | 4193046 | ||
| BC037230.1 | - | 1 | 209 | 2650923 | ||

Tbc1d10a
Genomic Map