siRNA Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029636.3 | NP_083912.2 | 6, 7 | 854 | ||
| GenBank mRNA | AF456461.1 | - | 6, 7 | 885 | ||
| AK005416.1 | - | 2, 3 | 675 | |||
| AY014776.1 | - | 6, 7 | 854 | |||
| BC099415.1 | - | 6, 7 | 861 | 30786803 | ||

Ctsq
Genomic Map