siRNA Details
| Gene Symbol: Sbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145587.2 | NP_663562.1 | 4 | 1022 | ||
| GenBank mRNA | AK200867.1 | - | 2 | 190 | ||
| BC024114.1 | - | 4 | 763 | 5098034 | ||
| BC025837.1 | - | 4 | 859 | 5135723 | ||
| BC031759.1 | - | 4 | 1022 | 4194152 | ||

Sbk1
Genomic Map