siRNA Details
| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028821.3 | NP_083097.2 | 3 | 199 | ||
| GenBank mRNA | AK005826.1 | - | 3 | 285 | ||
| BC070454.1 | - | 156 | ||||
| BC125392.1 | - | 2 | 181 | 40129907 | ||
| BC125394.1 | - | 2 | 177 | 40129909 | ||

Dnal1
Genomic Map