siRNA Details
| Gene Symbol: Nhlrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175340.4 | NP_780549.1 | 1 | 1227 | ||
| GenBank mRNA | AK045746.1 | - | 1 | 1198 | ||
| BC062644.1 | - | 1 | 1196 | |||
| BC118951.1 | - | 1 | 1198 | 40093176 | ||
| BC118952.1 | - | 1 | 1196 | 40093177 | ||

Nhlrc1
Genomic Map