siRNA Details
| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177992.2 | NP_818773.1 | 3, 4 | 363 | ||
| GenBank mRNA | AK031004.1 | - | 3, 4 | 365 | ||
| AK075772.1 | - | 3, 4 | 321 | |||
| BC003886.1 | - | 3, 4 | 322 | 3590411 | ||
| BC024109.1 | - | 3, 4 | 319 | 5149353 | ||

Gmpr2
Genomic Map