siRNA Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 9 | 1268 | ||
| GenBank mRNA | BB621791.1 | - | 5 | 401 | ||
| BC006648.1 | - | 3 | 1931 | |||
| BC024115.1 | - | 9 | 1256 | 5100379 | ||
| BC033365.1 | - | 7 | 1698 | 5367653 | ||
| DV645139.1 | - | 1 | 76 | |||

Chmp7
Genomic Map