siRNA Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 9, 10 | 1372 | ||
| GenBank mRNA | BB621791.1 | - | 5, 6 | 505 | ||
| BC006648.1 | - | 3, 4 | 2035 | |||
| BC024115.1 | - | 9, 10 | 1360 | 5100379 | ||
| BC033365.1 | - | 7, 8 | 1802 | 5367653 | ||
| DV645139.1 | - | 1, 2 | 180 | |||

Chmp7
Genomic Map