siRNA Details
| Gene Symbol: Pkdcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134117.2 | NP_598878.2 | 3 | 1191 | ||
| GenBank mRNA | AB381893.1 | - | 3 | 1183 | ||
| AB436780.1 | - | 3 | 1172 | |||
| BC022157.1 | - | 2 | 338 | |||
| BC086639.1 | - | 4 | 1029 | 6828629 | ||

Pkdcc
Genomic Map