siRNA Details
| Gene Symbol: Pkdcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134117.2 | NP_598878.2 | 5 | 1349 | ||
| GenBank mRNA | AB381893.1 | - | 5 | 1341 | ||
| AB436780.1 | - | 5 | 1330 | |||
| AK133913.1 | - | 1 | 26 | |||
| BC022157.1 | - | 4 | 496 | |||
| BC086639.1 | - | 6 | 1187 | 6828629 | ||

Pkdcc
Genomic Map