siRNA Details
| Gene Symbol: Camk2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025438.2 | NP_001020609.1 | 17 | 1782 | ||
| NM_001025439.2 | NP_001020610.1 | 19 | 1884 | |||
| NM_001293663.1 | NP_001280592.1 | 19 | 1884 | |||
| NM_001293664.1 | NP_001280593.1 | 18 | 1824 | |||
| NM_001293665.1 | NP_001280594.1 | 18 | 1775 | |||
| NM_023813.4 | NP_076302.1 | 17 | 1782 | |||
| GenBank mRNA | AK012702.1 | - | 17 | 1403 | ||
| AK039076.1 | - | 19 | 1493 | |||
| AK132185.1 | - | 4 | 301 | |||
| AK142435.1 | - | 17 | 1725 | |||
| AK148840.1 | - | 18 | 1531 | |||
| AK190737.1 | - | 1 | 41 | |||
| AK220516.1 | - | 19 | 1884 | |||
| BC042895.1 | - | 17 | 2323 | |||
| BC057103.1 | - | 18 | 1691 | |||

Camk2d
Genomic Map