siRNA Details
| Gene Symbol: Napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025898.3 | NP_080174.1 | 8, 9 | 705 | ||
| GenBank mRNA | AK004826.1 | - | 8, 9 | 719 | ||
| AK005372.1 | - | 8, 9 | 707 | |||
| AK013895.1 | - | 8, 9 | 724 | |||
| AK050623.1 | - | 8, 9 | 769 | |||
| AK136689.1 | - | 8, 9 | 708 | |||
| AK171998.1 | - | 8, 9 | 769 | |||
| BC004804.1 | - | 8, 9 | 736 | 3584869 | ||

Napa
Genomic Map