siRNA Details
| Gene Symbol: Fam207a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133998.3 | NP_598759.1 | 2 | 305 | ||
| GenBank mRNA | AF391115.1 | - | 2 | 278 | ||
| AK076516.1 | - | 2 | 276 | |||
| AK088306.1 | - | 2 | 273 | |||
| AK160157.1 | - | 2 | 176 | |||
| BB840302.1 | - | 2 | 306 | |||
| BC034216.1 | - | 2 | 251 | 5008732 | ||

Fam207a
Genomic Map