siRNA Details
| Gene Symbol: Fam212b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163356.1 | NP_001156828.1 | 2 | 643 | ||
| NM_175398.4 | NP_780607.2 | 2 | 790 | |||
| GenBank mRNA | AK033558.1 | - | 2 | 723 | ||
| AK078368.1 | - | 2 | 643 | |||
| BC052371.1 | - | 2 | 723 | 6491043 | ||

Fam212b
Genomic Map