siRNA Details
| Gene Symbol: Ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180600.3 | NP_850931.2 | 6, 7 | 958 | ||
| GenBank mRNA | AK076148.1 | - | 6, 7 | 821 | ||
| AK083216.1 | - | 6, 7 | 951 | |||
| AK150776.1 | - | 6, 7 | 783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2q2| Gene Symbol: Ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180600.3 | NP_850931.2 | 6, 7 | 958 | ||
| GenBank mRNA | AK076148.1 | - | 6, 7 | 821 | ||
| AK083216.1 | - | 6, 7 | 951 | |||
| AK150776.1 | - | 6, 7 | 783 | |||