siRNA Details
Gene Symbol: Ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180600.3 | NP_850931.2 | 2, 3 | 563 | ||
GenBank mRNA | AK042515.1 | - | 2, 3 | 564 | ||
AK076148.1 | - | 2, 3 | 426 | |||
AK083216.1 | - | 2, 3 | 556 | |||
AK150776.1 | - | 2, 3 | 388 | |||
BC029111.1 | - | 2, 3 | 392 | 2631258 |
