siRNA Details
| Gene Symbol: Lncbate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080070.1 | - | 2 | 623 | ||
| DV056598.1 | - | 2 | 723 | |||
| DV056599.1 | - | 2 | 723 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lncbate1| Gene Symbol: Lncbate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080070.1 | - | 2 | 623 | ||
| DV056598.1 | - | 2 | 723 | |||
| DV056599.1 | - | 2 | 723 | |||