siRNA Details
| Gene Symbol: Akr7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025337.3 | NP_079613.3 | 5 | 792 | ||
| GenBank mRNA | AF525358.2 | - | 5 | 694 | ||
| AJ271800.1 | - | 5 | 709 | |||
| AJ271801.1 | - | 5 | 709 | |||
| AK002664.1 | - | 5 | 772 | |||
| AK003915.1 | - | 5 | 779 | |||
| AK143203.1 | - | 5 | 791 | |||
| AK179820.1 | - | 2 | 145 | |||
| BC031857.1 | - | 5 | 769 | 5124841 | ||

Akr7a5
Genomic Map