siRNA Details
| Gene Symbol: Krt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033073.3 | NP_149064.1 | 5 | 834 | ||
| GenBank mRNA | AF509888.1 | - | 5 | 811 | ||
| AK002434.1 | - | 5 | 812 | |||
| AK137217.1 | - | 812 | ||||
| AK146914.1 | - | 5 | 812 | |||
| BC010337.1 | - | 4 | 2828 | |||
| BC030403.1 | - | 5 | 845 | 5355662 | ||

Krt7
Genomic Map