siRNA Details
| Gene Symbol: Amhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144547.2 | NP_653130.2 | 3 | 452 | ||
| GenBank mRNA | AF503863.1 | - | 3 | 399 | ||
| BC119571.1 | - | 3 | 383 | 40057985 | ||
| BC119572.2 | - | 9 | 1413 | |||
| KC920727.1 | - | 2 | 188 | |||

Amhr2
Genomic Map