siRNA Details
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080639.3 | NP_542370.3 | 3 | 340 | ||
GenBank mRNA | AF282730.1 | - | 3 | 271 | ||
AK038820.1 | - | 3 | 339 | |||
AK040814.1 | - | 3 | 339 | |||
AK079347.1 | - | 3 | 341 | |||
AK140367.1 | - | 3 | 340 | |||
BC064046.1 | - | 3 | 320 | 875422 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
