siRNA Details
| Gene Symbol: Vmn1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053221.2 | NP_444451.2 | 1 | 993 | ||
| GenBank mRNA | BC125372.1 | - | 1 | 993 | 40129887 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r42| Gene Symbol: Vmn1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053221.2 | NP_444451.2 | 1 | 993 | ||
| GenBank mRNA | BC125372.1 | - | 1 | 993 | 40129887 | |