siRNA Details
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053246.3 | NP_444476.1 | 7, 8 | 1060 | ||
| GenBank mRNA | AF418207.1 | - | 6, 7 | 944 | ||
| AK032352.1 | - | 7, 8 | 1050 | |||
| AK043670.1 | - | 6, 7 | 972 | |||
| AK077612.1 | - | 7, 8 | 1052 | |||
| AK139363.1 | - | 6, 7 | 949 | |||
| AK142621.1 | - | 6, 7 | 967 | |||
| AK156485.1 | - | 7, 8 | 1060 | |||
| BC004705.1 | - | 7, 8 | 1021 | 3499419 | ||

Dok4
Genomic Map