siRNA Details
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053246.3 | NP_444476.1 | 6 | 899 | ||
| GenBank mRNA | AF418207.1 | - | 5 | 783 | ||
| AK032352.1 | - | 6 | 889 | |||
| AK043670.1 | - | 5 | 811 | |||
| AK077612.1 | - | 6 | 891 | |||
| AK139363.1 | - | 5 | 787 | |||
| AK142621.1 | - | 5 | 806 | |||
| AK156485.1 | - | 6 | 899 | |||
| BC004705.1 | - | 6 | 860 | 3499419 | ||

Dok4
Genomic Map