siRNA Details
Gene Symbol: Gpr182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007412.2 | NP_031438.2 | 2 | 329 | ||
GenBank mRNA | AK031480.1 | - | 264 | |||
BC125279.1 | - | 2 | 200 | |||
BY121440.1 | - | 2 | 330 | |||
D17292.1 | - | 2 | 224 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
