siRNA Details
| Gene Symbol: Adra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009633.3 | NP_033763.2 | 1 | 1244 | ||
| GenBank mRNA | AF332049.1 | - | 1 | 950 | ||
| AF332050.1 | - | 1 | 950 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adra2b| Gene Symbol: Adra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009633.3 | NP_033763.2 | 1 | 1244 | ||
| GenBank mRNA | AF332049.1 | - | 1 | 950 | ||
| AF332050.1 | - | 1 | 950 | |||
Set Membership: |
|