siRNA Details
| Gene Symbol: Crisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009638.3 | NP_033768.3 | 7, 8 | 673 | ||
| GenBank mRNA | AK009449.1 | - | 7, 8 | 674 | ||
| BC011150.1 | - | 7, 8 | 656 | 4165956 | ||
| L05559.1 | - | 7, 8 | 643 | |||
| M92849.1 | - | 7, 8 | 639 | |||

Crisp1
Genomic Map